Learn More
MSC Caplugs High Temperature Masking Tape, 7.5 mil Thick, 1 Roll
Catalog No. 5000677102
Encompass
This off-white, 7.5 mil thick high temperature masking tape features crepe paper and acrylic adhesive, designed for applications requiring clean removal and heat resistance up to 149°C (300°F).
  • Clean removal
  • Heat-resistant
  • Flake-resistant
  • High quality
Catalog No. 50-006-77102 Supplier MSC Supplier No. 33509357
This off-white, 7.5 mil thick high temperature masking tape features crepe paper and acrylic adhesive, designed for applications requiring clean removal and heat resistance up to 149°C (300°F).
  • Clean removal
  • Heat-resistant
  • Flake-resistant
  • High quality