Learn More
MSC Caplugs Masking Tape, High Temperature, 1 Roll
Catalog No. 5000802934
Encompass
This high-temperature masking tape is made of crepe paper with an acrylic adhesive, designed for applications requiring heat resistance and clean removal. It measures 2 in. wide and 60 yd. long, with a thickness of 7.5 mil.
  • Clean removal
  • Heat-resistant
  • Flake-resistant
  • High quality
  • Wet-strip application
Catalog No. 50-008-02934 Supplier MSC Supplier No. 48263214
This high-temperature masking tape is made of crepe paper with an acrylic adhesive, designed for applications requiring heat resistance and clean removal. It measures 2 in. wide and 60 yd. long, with a thickness of 7.5 mil.
  • Clean removal
  • Heat-resistant
  • Flake-resistant
  • High quality
  • Wet-strip application