Learn More
Bellco Glass, Inc. Capmat Clr Silicon 96W Round
SDP
Catalog No. 501945637
Encompass_Preferred
Capmat Clear Silicone 96-Well Round. Temperature stability from -85F-+400F. Easy puncturing for sample extraction, Resealable up to 200 punctures. PTFE Coated to resist effects of chemicals and organic solvents. Autoclavebale for sterilization. Extreme resistance to tearing. 10 per case.
Catalog No. 50-194-5637 Supplier Bellco Glass, Inc. Supplier No. 451120096
Capmat Clear Silicone 96-Well Round. Temperature stability from -85F-+400F. Easy puncturing for sample extraction, Resealable up to 200 punctures. PTFE Coated to resist effects of chemicals and organic solvents. Autoclavebale for sterilization. Extreme resistance to tearing. 10 per case.