Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One CapMat, for use with 96-well MASTERBLOCK™ Microplates
GREENER_CHOICE

Catalog No. 07000177 Shop All Greiner Bio-One Products
Change view
Click to view available options
Pre-slit:
No
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pre-slit
07-000-177 No
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-000-177 Supplier Greiner Bio-One Supplier No. 381061
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sealers for microplates

  • For use with 96-well MASTERBLOCK Microplates
TRUSTED_SUSTAINABILITY

Specifications

Color Translucent
Solvent Or Alcohol Resistant DMSO Resistant
Certifications/Compliance Free of detectable DNase/RNase, human DNA and pyrogens; resistant to DMSO; non-cytotoxic
Material EVA
Sterility Sterile
Autoclavable Not Autoclavable
No. of Wells 96
No. per Case 50
No. per Pack 1
Pre-slit No
Product Type Sealing Mat
Adhesive None
Certified RNase/DNase-free Yes
Pierceable No
Temperature Range (Metric) -60°C to +65°C
Well Shape Round
Plate Compatibility 96-well MASTERBLOCK (0.5 and 1 mL), Standard 96-well Polypropylene Microplates, 96-well F-, U-, and μ Clear-bottom Polystyrene Microplates
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.