Learn More
Medchemexpress LLC Capmatinib dihydrochloride | 1197376-85-4 | 99.8% | C23H19Cl2FN6O | 10MG
SDP
Supplier:  Medchemexpress LLC HY13404A10MG
Encompass_Preferred
Capmatinib dihydrochloride is the dihydrochloride salt of capmatinib, a potent, selective, ATP-competitive inhibitor of the c-Met (HGFR) tyrosine kinase. Supplied as a research-grade solid, it is intended for in vitro and preclinical oncology studies and is accompanied by supplier analytical data documenting high purity and activity.
  • Selective, ATP-competitive c-Met (HGFR) kinase inhibitor.
  • Very high in vitro potency (IC50 = 0.13 nM against c-Met).
  • High purity suitable for research (reported ~99.8%).
  • Suitable for in vitro and preclinical oncology assays.
  • Available as small-scale solid quantities for laboratory use.
Catalog No. 50-002-60390