Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Capmatinib dihydrochloride | 1197376-85-4 | MFCD30502991 | 99.8% | 485.34 g/mol | C23H19Cl2FN6O | 5 MG
SDP

Catalog No. 5000262287
Encompass_Preferred

Capmatinib dihydrochloride is the dihydrochloride salt of capmatinib, a potent, selective, ATP-competitive inhibitor of the c-MET tyrosine kinase for preclinical research. It inhibits c-MET phosphorylation and downstream effectors, suppressing c-MET-dependent tumor cell proliferation and migration.

  • Potent c-MET kinase inhibitor with low nanomolar activity.
  • Inhibits downstream signaling pathways involved in proliferation and survival.
  • Supplied as a dihydrochloride salt to aid solubility for assays.
  • High purity (99.8%) suitable for analytical and biological studies.
  • Available in small pack sizes for laboratory research use.
  • Molecular weight 485.34 g/mol; formula C23H19Cl2FN6O.

Catalog No. 50-002-62287 Supplier Medchemexpress LLC Supplier No. HY13404A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Capmatinib dihydrochloride is the dihydrochloride salt of capmatinib, a potent, selective, ATP-competitive inhibitor of the c-MET tyrosine kinase for preclinical research. It inhibits c-MET phosphorylation and downstream effectors, suppressing c-MET-dependent tumor cell proliferation and migration.

  • Potent c-MET kinase inhibitor with low nanomolar activity.
  • Inhibits downstream signaling pathways involved in proliferation and survival.
  • Supplied as a dihydrochloride salt to aid solubility for assays.
  • High purity (99.8%) suitable for analytical and biological studies.
  • Available in small pack sizes for laboratory research use.
  • Molecular weight 485.34 g/mol; formula C23H19Cl2FN6O.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.