Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Capmatinib dihydrochloride hydrate | 1865733-40-9 | 99.8% | 503.36 g·mol-1 | C23H21Cl2FN6O2 | 5 MG
SDP

Catalog No. 5000262535
Encompass_Preferred

Capmatinib dihydrochloride hydrate is the dihydrochloride hydrate salt form of capmatinib, a selective, orally active c-Met kinase inhibitor used in research to investigate MET-driven signaling and anticancer activity. Supplied as a high-purity solid for laboratory use.

  • Selective c-Met kinase inhibitor suitable for MET-related studies.
  • High purity (≈99.8%).
  • Molecular weight 503.36 g·mol-1; formula C23H21Cl2FN6O2.
  • White to light yellow solid physical form.
  • Provided as a research-grade chemical for in vitro and in vivo assays.
  • Store sealed, away from moisture and light; follow low-temperature solvent storage guidance.

Catalog No. 50-002-62535 Supplier Medchemexpress LLC Supplier No. HY13404C5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Capmatinib dihydrochloride hydrate is the dihydrochloride hydrate salt form of capmatinib, a selective, orally active c-Met kinase inhibitor used in research to investigate MET-driven signaling and anticancer activity. Supplied as a high-purity solid for laboratory use.

  • Selective c-Met kinase inhibitor suitable for MET-related studies.
  • High purity (≈99.8%).
  • Molecular weight 503.36 g·mol-1; formula C23H21Cl2FN6O2.
  • White to light yellow solid physical form.
  • Provided as a research-grade chemical for in vitro and in vivo assays.
  • Store sealed, away from moisture and light; follow low-temperature solvent storage guidance.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.