Learn More
Selleck Chemical LLC Capmatinib (INCB28060) 10mg 1029712-80-8 INC280, NVP-INC280
SDP
Supplier:  Selleck Chemical LLC S278810MG
Encompass_Preferred
Capmatinib (INCB28060, INC280, NVP-INC280) is a novel, ATP-competitive inhibitor of c-MET with IC50 of 0.13 nM in a cell-free assay, inactive against RON?, as well as EGFR and HER-3. Capmatinib (INCB28060) inhibits Wnt/?-catenin and EMT signaling pathways and induces apoptosis in diffuse gastric cancer positive for c-MET amplification. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-7965