Learn More
Aviva Systems Biology Corp CAPN1 Antibody - N-terminal region (ARP60937_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60937_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CAPN1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human CAPN1. Alternate names: CANP, CANP1, CANPL1, muCANP, muCLApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-287-206