Learn More
Aviva Systems Biology Corp CAPN1 Antibody - middle region (ARP60936_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60936_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CAPN1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human CAPN1. Alternate names: CANP, CANP1, CANPL1, muCANP, muCLApplicationWB, IHC. Species reactivity: Cow, Goat, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-287-205