Learn More
Aviva Systems Biology Corp CAPN3 Antibody - C-terminal region (ARP60946_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP60946_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CAPN3. It was validated on Western Blot. . Alternate names: CANP3, CANPL3, LGMD2, LGMD2A, MGC10767, MGC11121, MGC14344, MGC4403, nCL-1, p94ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Rabbit, Rat, Sheep
Catalog No. 50-287-207