Learn More
Med Vet International Micro Capnometer, J0166Q, Each
Catalog No. 5000495828
Encompass_Preferred
The smallest main stream capnograph available and only weighs 50 gms. • No calibration needed • Large display with capnograph wave form • Numerical respiratory rate • Alarms • Insert between endotracheal tube and anesthesia circuit • Long battery life, up to 10 hours normal use with two AAA lithium batteries
Catalog No. 50-004-95828 Supplier Med Vet International Supplier No. JOR166Q
The smallest main stream capnograph available and only weighs 50 gms. • No calibration needed • Large display with capnograph wave form • Numerical respiratory rate • Alarms • Insert between endotracheal tube and anesthesia circuit • Long battery life, up to 10 hours normal use with two AAA lithium batteries