Learn More
Marketlab CapnoVue™ SCOPE Face Mask
Description
- Allows for easy, reliable, and consistent capnography during transoral sedation procedures
- Minimizes friction and procedural interference
- Large 20 mm aperture accommodates most instruments and bite blocks
- Fast, easy sample-line attachment with visual confirmation of connections
- Intuitive EtCO2 sample line assembly
- Patented, integrated female luer lock sampling ports are ideally situated on the mask to provide reliable EtCO2 sampling
- Optimally placed sampling ports in the gas exhalation vents provide consistent readings for both nose and mouth breathing
Specifications
Specifications
| Product Type | Oxygen Mask |
| Color | Clear |
| Material | PVC |
| Sterility | Non-sterile |