Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Capping A Reagent for AKTA Oligopilot-100, NovaBiochem™ MilliporeSigma™

Catalog No. MBI02244000 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MBI02244000 4 L
MBI02241005 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MBI02244000 Supplier MilliporeSigma Supplier No. BI02244000
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • Used for analysis
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Methylimidazole/Acetonitrile (20/80, v/v)
CAS 616-47-7, 75-05-8
Density 0.787g/cm3 (at 20°C)
Flash Point 5.5°C
Odor Solvent odor
Packaging Glass bottle (pre-packed, 38/430)
Molecular Formula CH3-C3H3N2 in CH3CN
Quantity 4 L
Synonym DNA
Formula Weight Mixture
Percent Purity 79.0 to 81.0% (v/v)
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.