Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Capping Reagent B,16%, n-Methylimidazole in THF, Novabiochem™ MilliporeSigma™

Catalog No. MBI03454000 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
4 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MBI03454000 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MBI03454000 Supplier MilliporeSigma Supplier No. BI03454000
Add to Cart
Add to Cart

Chemical Identifiers

Molecular Formula Mixture

Specifications

Melting Point -108.5°C
Boiling Point 65°C to 66°C (1013 hPa)
Flash Point −21.5°C
Packaging Glass Bottle
Quantity 4 L
Vapor Pressure 173 hPa (20°C)
Percent Purity 15.6-16.4%
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.