Learn More
Sigma Aldrich Fine Chemicals Biosciences n-Methylimidazole | Purity: 16% | CAS No: 616-47-7 | 4 L
Catalog No. NC2469000
Encompass
n-Methylimidazole | Purity: 16% | Mol Wt: 82.11 | CAS No: 616-47-7 | Unit of Measure: 4LIn tetrahydrofuran (THF) solvent for capping unreacted 5'-hydroxyl groups in DNA/RNA synthesis to prevent side reactions and improve yield. Classified as a flammable liquid slightly hazardous to water (WGK 1). Also known as Novabiochem Capping Reagent B. Packaged in 38/430 glass bottles.
Catalog No. NC2469000 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. BI03454000
n-Methylimidazole | Purity: 16% | Mol Wt: 82.11 | CAS No: 616-47-7 | Unit of Measure: 4LIn tetrahydrofuran (THF) solvent for capping unreacted 5'-hydroxyl groups in DNA/RNA synthesis to prevent side reactions and improve yield. Classified as a flammable liquid slightly hazardous to water (WGK 1). Also known as Novabiochem Capping Reagent B. Packaged in 38/430 glass bottles.