Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Capping Reagent B1 (40% Acetic Anhydride in Acetonitrile), MilliporeSigma™

Catalog No. MBI03474000 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
4 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MBI03474000 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MBI03474000 Supplier MilliporeSigma Supplier No. BI03474000
Add to Cart
Add to Cart
  • Used for biosynthesis processes such as DNA synthesis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula Mixture
Synonym Capping reagent

Specifications

Density 0.9 g/cm3
Quantity 4 L
Flash Point 9.4°C
Packaging Glass Bottle
Physical Form Liquid
Water 100 ppm
UN Number UN 2924
Formula Weight Mixture
Percent Purity 99.0% (min)
Hazard Category Danger
Hazard Statement H225: Highly flammable liquid and vapor, H302 + H312: Harmful if swallowed or in contact with skin, H314: Causes severe skin burns and eye damage, H330: Fatal if inhaled, H335: May cause respiratory irritation, H335: May cause respiratory irritation.
Precautionary Statement P210: Keep away from heat/sparks/open flames/hot surfaces. No smoking, P233: Keep container tightly closed, P240: Ground/bond container and receiving equipment, P241: Use explosion-proof electrical/ ventilating/ lighting/ equipment, P242: Use only non-spa
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.