Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Capping Reagent, B2 60%, Lutidine in Acetonitrile, Novabiochem™, MilliporeSigma™

Catalog No. MBI03490505 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MBI03490505 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MBI03490505 Supplier MilliporeSigma Supplier No. BI03490505
Add to Cart
Add to Cart
This item is not returnable. View return policy

For DNA synthesis

Recommended Storage

Store below +30°C.

TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Format Liquid
Packaging Glass Bottle
Quantity 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.