Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Capping Reagent, Solstice

Catalog No. 60090055 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
56 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-090-055 56 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-090-055 Supplier Solstice Supplier No. BC73356
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

BioSyn™, suitable for use with DNA/RNA synthesis instruments, 50/30/20 Acetonitrile/Lutidine/NMI (v/v)

TRUSTED_SUSTAINABILITY

Specifications

CAS 616-47-7, 108-48-5, 75-05-8
CAS Max % 23.60%, 31.60%, 44.80%
Quantity 56 L
Flash Point 10°C
Packaging Returnable Steel Drum
Physical Form Liquid, clear
Purity Grade Notes Capping reagents
Boiling Point 73.89°C
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.