Learn More
Solstice Capping Reagent Kit, contains 40% Acetic Anhydride/ 60% Acetonitrile (BR643) and 60% 2,6-Lutidine / 40%Acetonitrile (BR645)
Shop All Solstice Research Chemicals ProductsDescription
Solstice BioSyn™ reagents and solvents for DNA/RNA synthesis
- Solstice BioSyn™ Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
- ideal for oligonucleotide synthesis regardless of the application
- All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination
Specifications
Specifications
| Content And Storage | Room Temperature |
| Packaging Type | Glass Bottle |
| Quantity | 1 L Kit |
| Form | Liquid |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.