Learn More
Capping Reagent Kit, Solstice
Catalog No. 60090163 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
5 L Kit
Catalog No. Quantity
60-090-163 5 L Kit
1 options
Catalog No. 60-090-163 Supplier Solstice Supplier No. BR6482.5
Add to Cart
Add to Cart
BioSyn™, suitable for use with DNA/RNA synthesis instruments, contains 40% Acetic Anhydride/ 60% BioSyn™, suitable for use with DNA/RNA synthesis instruments, Acetonitrile (BR643) and 60% 2,6-Lutidine / 40%Acetonitrile (BR645)
TRUSTED_SUSTAINABILITY
Specifications
Quantity 5 L Kit
Packaging Glass Bottle
Purity Grade Notes Capping reagents