Learn More
Pyramex™ Cappture™ Safety Eyewear
Catalog No. 19155630 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Lens Tint:
Clear
Gray
Lens Type:
H2MAX
H2X
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint Lens Type
19-155-630 Gray H2X
19-155-629 Clear H2X
19-151-529 Clear H2MAX
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-155-630 Supplier Pyramex™ Supplier No. S9920ST
Add to Cart
Add to Cart
This item is not returnable. View return policy
Cappture™ Safety Eyewear fits over your prescription frames.
  • Dielectric
  • Temples extend from browbar to avoid interference with prescription frame temples
Specifications
Certifications/Compliance ANSI Z87.1, CSA Z94.3
Lens Coating Anti-fog, UV-treated, Scratch-resistant, Impact-resistant
Lens Type H2X
Certifications/Compliance (Europe) EN-166
Lens Tint Gray
Product Type Wrap-around Safety Glasses
For Use With (Application) Fit over prescription glasses
Material Polycarbonate
Disposable Reusable
Lens Material Polycarbonate
Frame Material Polycarbonate
UV Protection U6
Frame Type Frameless Uni-lens
Autoclavable Not Autoclavable