Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Capreomycin (sulfate) | 1405-37-4 | 99.5% | 1 G
SDP

Catalog No. 5000386919
Encompass_Preferred

Capreomycin sulfate is a macrocyclic peptide antibiotic that inhibits phenylalanine synthesis in mycobacterial ribosomal translation. It exhibits anti-amyloidogenic and pro-fibrinolytic activities, reducing amyloid-induced cytotoxicity by inhibiting amyloid fibrillation. This product is suitable for studies related to multidrug-resistant tuberculosis, type 2 diabetes, Alzheimer's disease, and Parkinson's disease.

  • Inhibits insulin fibril formation in human erythrocytes.
  • Protects against amyloid fibril-induced erythrocyte damage and hemolysis.
  • Destroys fibrils in erythrocytes in a time-dependent manner.
  • Decreases wet lung weight and bacterial burden in the lungs of infected guinea pigs.
  • Appearance: solid.
  • Color: white to off-white.
  • Initial source: microorganisms (Streptomyces capreolus).
  • Storage: -20°C, sealed, away from moisture.
  • Solubility in H2O: 100 mg/mL (requires ultrasonic).

Catalog No. 50-003-86919 Supplier Medchemexpress LLC Supplier No. HY175661G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Capreomycin sulfate is a macrocyclic peptide antibiotic that inhibits phenylalanine synthesis in mycobacterial ribosomal translation. It exhibits anti-amyloidogenic and pro-fibrinolytic activities, reducing amyloid-induced cytotoxicity by inhibiting amyloid fibrillation. This product is suitable for studies related to multidrug-resistant tuberculosis, type 2 diabetes, Alzheimer's disease, and Parkinson's disease.

  • Inhibits insulin fibril formation in human erythrocytes.
  • Protects against amyloid fibril-induced erythrocyte damage and hemolysis.
  • Destroys fibrils in erythrocytes in a time-dependent manner.
  • Decreases wet lung weight and bacterial burden in the lungs of infected guinea pigs.
  • Appearance: solid.
  • Color: white to off-white.
  • Initial source: microorganisms (Streptomyces capreolus).
  • Storage: -20°C, sealed, away from moisture.
  • Solubility in H2O: 100 mg/mL (requires ultrasonic).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.