Learn More
TARGETMOL CHEMICALS INC Capreomycin sulfate 1G
SDP
Catalog No. 502880497
Encompass_Preferred
Also available in 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Capreomycin sulfate (Capastat sulfate) , a cyclic peptide antibiotic, binds to the 70S ribosomal unit leading to inhibition of protein synthesis. Purity 99.54%
Catalog No. 50-288-0497 Supplier TARGETMOL CHEMICALS INC Supplier No. T14411G
Also available in 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Capreomycin sulfate (Capastat sulfate) , a cyclic peptide antibiotic, binds to the 70S ribosomal unit leading to inhibition of protein synthesis. Purity 99.54%