Learn More
Apexbio Technology LLC Capreomycin Sulfate 1405-37-4 1g
SDP
Catalog No. 502240596
Encompass_Preferred
Capreomycin sulfate (CAS 1405-37-4) is a cyclic peptide antibiotic targeting the bacterial 70S ribosomal subunit It is designed to disrupt translation processes thereby inhibiting bacterial protein biosynthesis Capreomycin sulfate exerts its biological activity primarily through specific binding to the ribosomal subunit inhibiting protein synthesis In in vitro studies Capreomycin sulfate demonstrates inhibitory activity with reported IC50 values typically ranging from approximately 2 to 10 g/mL dependent upon the bacterial species and experimental conditions Based on these pharmacological properties Capreomycin sulfate holds research potential in antimicrobial susceptibility testing and the study of resistance mechanisms in Mycobacterium tuberculosis strains
This item is not returnable. View return policy
Catalog No. 50-224-0596 Supplier Apexbio Technology LLC Supplier No. B16891000
Capreomycin sulfate (CAS 1405-37-4) is a cyclic peptide antibiotic targeting the bacterial 70S ribosomal subunit It is designed to disrupt translation processes thereby inhibiting bacterial protein biosynthesis Capreomycin sulfate exerts its biological activity primarily through specific binding to the ribosomal subunit inhibiting protein synthesis In in vitro studies Capreomycin sulfate demonstrates inhibitory activity with reported IC50 values typically ranging from approximately 2 to 10 g/mL dependent upon the bacterial species and experimental conditions Based on these pharmacological properties Capreomycin sulfate holds research potential in antimicrobial susceptibility testing and the study of resistance mechanisms in Mycobacterium tuberculosis strains