Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Capreomycin sulfate, 90%

Catalog No. AAJ66684MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ66684MD 250 mg
AAJ6668403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ66684MD Supplier Thermo Scientific Chemicals Supplier No. J66684MD
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1405-37-4 | C25H46N14O12S | 766.79 g/mol

Thought to inhibit protein synthesis by inhibiting normal ribosomal functions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Thought to inhibit protein synthesis by inhibiting normal ribosomal functions
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1405-37-4
Molecular Formula C25H46N14O12S
Molecular Weight (g/mol) 766.79
MDL Number MFCD00079032
InChI Key AJQVUIHGEOLMDY-SOCRLDLMNA-N
IUPAC Name 3,6-diamino-N-{[(8Z)-15-amino-11-(2-amino-3,4,5,6-tetrahydropyrimidin-4-yl)-8-[(carbamoylamino)methylidene]-2-(hydroxymethyl)-3,6,9,12,16-pentaoxo-1,4,7,10,13-pentaazacyclohexadecan-5-yl]methyl}hexanamide; sulfuric acid
SMILES OS(O)(=O)=O.NCCCC(N)CC(=O)NCC1NC(=O)C(CO)NC(=O)C(N)CNC(=O)C(NC(=O)\C(NC1=O)=C\NC(N)=O)C1CCN=C(N)N1

Specifications

Type Capreomycin Sulfate
Physical Form Powder
Color White
Quantity 250 mg
Flash Point >110°C (230°F)
Boiling Point 1377°C
Solubility Information Freely soluble in water
Formula Weight 766.78
Percent Purity 90%
Chemical Name or Material Capreomycin sulfate
Hazard Category H302+H312+H332-H360
Hazard Statement GHS H Statement
H360-H302-H312-H332
May damage fertility or the unborn child.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P308+P313-P312-P330-P363-P501c
EINECSNumber 215-776-8
RTECSNumber EX8930000
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.