Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Capreomycin Sulfate, MP Biomedicals™

Catalog No. MP215492491 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215492491 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP215492491 Supplier MP Biomedicals™ Supplier No. 0215492491
Add to Cart
Add to Cart

Often grouped with aminoglycosides. Capreomycin Sulfate, MP Biomedicals is a cyclic peptide antibiotic. This second line antibiotic binds across the ribosomal interface involving 23S rRNA helix 69 (H69) and 16S rRNA helix 44 (h44).

Capreomycin is used to study multidrug-resistant tuberculosis. It is also used to study bacterial ribosomal subunit interactions and translocation processes during protein synthesis.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1405-37-4
Content And Storage -20°C
Format Powder
For Use With (Application) Capreomycin is a second line antibiotic and is used to study multidrug-resistant tuberculosis. Capreomycin sulfate is used to study bacterial ribosomal subunit interactions and translocation processes during protein synthesis.
Molecular Weight (g/mol) 750.79
Quantity 1 g
Source Streptomyces capreolus
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.