Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Capreomycin Sulfate MP Biomedicals

Catalog No. ICN15492483 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15492483 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15492483 Supplier MP Biomedicals, Inc Supplier No. 0215492483
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1405-37-4
Molecular Formula C25H46N14O12S
Molecular Weight (g/mol) 766.79
MDL Number MFCD00079032
InChI Key AJQVUIHGEOLMDY-SOCRLDLMNA-N
Synonym Capastal sulfate, Caprocin
IUPAC Name 3,6-diamino-N-{[(8Z)-15-amino-11-(2-amino-3,4,5,6-tetrahydropyrimidin-4-yl)-8-[(carbamoylamino)methylidene]-2-(hydroxymethyl)-3,6,9,12,16-pentaoxo-1,4,7,10,13-pentaazacyclohexadecan-5-yl]methyl}hexanamide; sulfuric acid
SMILES OS(O)(=O)=O.NCCCC(N)CC(=O)NCC1NC(=O)C(CO)NC(=O)C(N)CNC(=O)C(NC(=O)\C(NC1=O)=C\NC(N)=O)C1CCN=C(N)N1

Specifications

Type Capreomycin Sulfate
Quantity 250 mg
Formula Weight 750.786
Chemical Name or Material Capreomycin Sulfate
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.