Learn More
Selleck Chemical LLC Capric acid
SDP
Supplier:  Selleck Chemical LLC S6906100MG
Encompass_Preferred
Capric acid (DA Decanoic acid Decylic acid) a component of medium-chain triclycerides occurring naturally in coconut oil and palm kernel oil contributes to seizure control through direct AMPA receptor inhibition Capric acid is also a modulating ligand for PPARs Due to its specific melting range the product may be solid liquid a solidified melt or a supercooled melt
Catalog No. 50-000-02295