Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC ε-caprolactone-d6 | 1219802-08-0 | 99.3% | 120.18 g·mol^-1 | C6H4D6O2 | 5 MG
SDP

Catalog No. 5000299929
Encompass_Preferred

Deuterium-labeled ε-caprolactone is a stable isotope reagent used as a tracer and internal standard for quantitative NMR, GC-MS, and LC-MS. It is supplied in small laboratory-scale quantities and is characterized by the deuterated formula C6H4D6O2 and a molecular weight of 120.18 g·mol⁻1.

  • High chemical purity: 99.3%.
  • Suitable as tracer and internal standard for quantitative analysis.
  • Deuterated molecular formula: C6H4D6O2.
  • Appearance: colorless to light yellow liquid.
  • Long-term storage: -20°C for pure material; in solvent store at -80°C (6 months) or -20°C (1 month).
  • Available in small milligram pack sizes for research use.

Catalog No. 50-002-99929 Supplier Medchemexpress LLC Supplier No. HYW015891S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Deuterium-labeled ε-caprolactone is a stable isotope reagent used as a tracer and internal standard for quantitative NMR, GC-MS, and LC-MS. It is supplied in small laboratory-scale quantities and is characterized by the deuterated formula C6H4D6O2 and a molecular weight of 120.18 g·mol⁻1.

  • High chemical purity: 99.3%.
  • Suitable as tracer and internal standard for quantitative analysis.
  • Deuterated molecular formula: C6H4D6O2.
  • Appearance: colorless to light yellow liquid.
  • Long-term storage: -20°C for pure material; in solvent store at -80°C (6 months) or -20°C (1 month).
  • Available in small milligram pack sizes for research use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.