Learn More
Medchemexpress LLC ε-caprolactone-d6 | 1219802-08-0 | MFCD00003267 | 99.3% | 120.18 g/mol | C6H4D6O2 | 1 MG
SDP
Catalog No. 5000300558
Encompass_Preferred
ε-Caprolactone-d6 is the deuterium-labeled isotopologue of ε-caprolactone (C6H4D6O2, CAS 1219802-08-0). Supplied as a high-purity liquid, it is intended for use as an isotopic tracer and internal standard in analytical and quantitative chemistry applications, with supplier guidance for storage and handling to maintain stability.
  • Deuterium-labeled internal standard for quantitative analysis.
  • Suitable for NMR, GC-MS, and LC-MS applications.
  • High reported purity of 99.3%.
  • Liquid, colorless to light yellow physical form for easy handling.
  • Storage guidance provided to preserve stability (e.g., -20°C for pure material).
Catalog No. 50-003-00558 Supplier Medchemexpress LLC Supplier No. HYW015891S1MG
ε-Caprolactone-d6 is the deuterium-labeled isotopologue of ε-caprolactone (C6H4D6O2, CAS 1219802-08-0). Supplied as a high-purity liquid, it is intended for use as an isotopic tracer and internal standard in analytical and quantitative chemistry applications, with supplier guidance for storage and handling to maintain stability.
  • Deuterium-labeled internal standard for quantitative analysis.
  • Suitable for NMR, GC-MS, and LC-MS applications.
  • High reported purity of 99.3%.
  • Liquid, colorless to light yellow physical form for easy handling.
  • Storage guidance provided to preserve stability (e.g., -20°C for pure material).