Learn More
Medchemexpress LLC HY-15243 5mg Medchemexpress, Capromorelin (Tartrate) CAS:193273-69-7 Purity:>98%
SDP
Catalog No. 502081695
Encompass_Preferred
Medchemexpress, HY-15243 5mg Capromorelin (Tartrate) CAS:193273-69-7 Capromorelin Tartrate is an orally active, potent growth hormone secretagogue receptor (GHSR) agonist, with Ki of 7 nM for hGHS-R1a. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1695 Supplier Medchemexpress LLC Supplier No. HY152435MG
Medchemexpress, HY-15243 5mg Capromorelin (Tartrate) CAS:193273-69-7 Capromorelin Tartrate is an orally active, potent growth hormone secretagogue receptor (GHSR) agonist, with Ki of 7 nM for hGHS-R1a. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.