Learn More
Cayman Chemical CapromorelIntartrate 10mg
SDP
Catalog No. 502950836
Encompass_Preferred
A GH secretagogue and an agonist of GHS-R1a; stimulates GH release from primary rat pituitary cells EC50 = 3 nM); stimulates intracellular calcium mobilization in primary rat pituitary cells at 0.3, 3, and 30 nM; increases rat serum GH levels (ED50 = 0.04 mg/kg, i.v.); decreases dog serum IGF-1 levels at 1 mg/kg, p.o.; increases rat body weight by 43.6% without affecting whole-body percentages of fat, lean mass, or bone content at 3.9 mg/kg, once per day for 28 days; decreases infarct size in a rabbit model of ischemia-reperfusion injury induced by left coronary artery occlusion at 25 mg/kg
Catalog No. 50-295-0836 Supplier Cayman Chemical Supplier No. 3348810 MG
A GH secretagogue and an agonist of GHS-R1a; stimulates GH release from primary rat pituitary cells EC50 = 3 nM); stimulates intracellular calcium mobilization in primary rat pituitary cells at 0.3, 3, and 30 nM; increases rat serum GH levels (ED50 = 0.04 mg/kg, i.v.); decreases dog serum IGF-1 levels at 1 mg/kg, p.o.; increases rat body weight by 43.6% without affecting whole-body percentages of fat, lean mass, or bone content at 3.9 mg/kg, once per day for 28 days; decreases infarct size in a rabbit model of ischemia-reperfusion injury induced by left coronary artery occlusion at 25 mg/kg