Learn More
Cayman Chemical Caprooyl Tetrapeptide-3 (acetate), Purity: ≥98%, CAS: 1012317-71-3, 10 mg
SDP
Catalog No. 502937035
Encompass_Preferred
A derivative of tetrapeptide-3 linked to caproic acid; has migration-imparting effects in human adipose-derived stem cells when applied at concentrations of 0.1 and 1 mg/ml
Catalog No. 50-293-7035 Supplier Cayman Chemical Supplier No. 2715410 MG
A derivative of tetrapeptide-3 linked to caproic acid; has migration-imparting effects in human adipose-derived stem cells when applied at concentrations of 0.1 and 1 mg/ml