Learn More
Medchemexpress LLC HY-135087 100mg Medchemexpress, Caprylic/Capric Triglyceride CAS:65381-09-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY135087100MG
Encompass_Preferred
Medchemexpress, HY-135087 100mg Caprylic/Capric Triglyceride CAS:65381-09-1 Caprylic/Capric Triglyceride is the triglycerides and esters prepared from fractionated vegetable oil sources and fatty acids from coconuts and palm kernel oils. Caprylic/Capric Triglyceride possesses excellent oxidation stability. Caprylic/Capric Triglyceride is used as a food additive and used in cosmetics. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1377