Learn More
Apexbio Technology LLC Caprylic/Capric-Triglyceride
SDP
Catalog No. 5000568782
Encompass_Preferred
Caprylic/Capric Triglyceride (CAS No 65381-09-1) is a medium-chain triglyceride commonly used in biomedical research to investigate lipid metabolism and energy regulation pathways Originating from the esterification of glycerol with caprylic and capric fatty acids it modulates biological processes by providing a readily absorbable energy source In vitro its activity is often assessed within nanomolar to low micromolar concentrations to examine cellular uptake and metabolism dynamics Typically applied in cell models and animal studies it aids in drug screening and pharmacokinetic evaluations where it serves as a model compound to study lipid absorption and bioavailability Experimental concentrations typically depend on the specific research design allowing for flexible application across various disciplines including dermatology and neurology where it is valued for its stability and biocompatibility
Catalog No. 50-005-68782 Supplier Apexbio Technology LLC Supplier No. C6514100
Caprylic/Capric Triglyceride (CAS No 65381-09-1) is a medium-chain triglyceride commonly used in biomedical research to investigate lipid metabolism and energy regulation pathways Originating from the esterification of glycerol with caprylic and capric fatty acids it modulates biological processes by providing a readily absorbable energy source In vitro its activity is often assessed within nanomolar to low micromolar concentrations to examine cellular uptake and metabolism dynamics Typically applied in cell models and animal studies it aids in drug screening and pharmacokinetic evaluations where it serves as a model compound to study lipid absorption and bioavailability Experimental concentrations typically depend on the specific research design allowing for flexible application across various disciplines including dermatology and neurology where it is valued for its stability and biocompatibility