Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caprylic/Capric Triglyceride, Spectrum™ Chemical
SDP

Catalog No. 18620468 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-620-468 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-620-468 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. C3465500ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 73398-61-5

Specifications

Boiling Point 150°C
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Caprylic/Capric Triglyceride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.