Learn More
Research Products International Corp CAPS [3-(Cyclohexylamino)-1-propanesulphonic acid], 1 Kilograms
SDP
Catalog No. 501368152
Encompass_Preferred
pKa (25oC): 10.4. The working pH range in aqueous solution is 9.7-11.1
Catalog No. 50-136-8152 Supplier Research Products International Corp Supplier No. C300601000.0
pKa (25oC): 10.4. The working pH range in aqueous solution is 9.7-11.1