Learn More
Microsolv Technology Corporation Caps 10-425 Preslit Sil/Ptfec
Catalog No. 5001452201
Encompass_Preferred
Caps, Screw Top, with Silicone Rubber / PTFE Pre-Slit Septa, bonded in wide mouth, knurled, polypropylene, Black Caps. For use with 10-425mm threads, 12x32mm autosampler vials. BASIK Brand. 100/PK.
Catalog No. 50-014-52201 Supplier Microsolv Technology Corporation Supplier No. 9503S30CK
Caps, Screw Top, with Silicone Rubber / PTFE Pre-Slit Septa, bonded in wide mouth, knurled, polypropylene, Black Caps. For use with 10-425mm threads, 12x32mm autosampler vials. BASIK Brand. 100/PK.