Learn More
Research Products International Corp CAPS [3-(Cyclohexylamino)-1-propanesulphonic acid], 100 Grams
SDP
Catalog No. 50488577
Encompass_Preferred
pKa (25oC): 10.4. The working pH range in aqueous solution is 9.7-11.1
Catalog No. 50-488-577 Supplier Research Products International Corp Supplier No. C30060100.0
pKa (25oC): 10.4. The working pH range in aqueous solution is 9.7-11.1