Learn More
Andwin Scientific CAPS 1ML/MILES NO 4335 200/PK
SDP
Catalog No. NC9436586
Encompass_Preferred
The 1 mL Chamber caps is an accessory for the Cyto-Tek Centrifuge Funnel Chamber, which is a product made to provide cell recovery up to 100%. The caps are sold separately in a pack of 200.
Catalog No. NC9436586 Supplier Andwin Scientific Supplier No. C16607
The 1 mL Chamber caps is an accessory for the Cyto-Tek Centrifuge Funnel Chamber, which is a product made to provide cell recovery up to 100%. The caps are sold separately in a pack of 200.