Learn More
Microsolv Technology Corporation Caps 2Ml Nat Slit Sil/Ptfe Bl
Catalog No. 5001452132
Encompass_Preferred
Caps, Screw Top, with Natural Silicone Rubber / White PTFE Pre-Slit Septa, bonded in knurled, polypropylene, Blue Caps. For use with 9-425mm thread, 12x32mm autosampler vials. MicroSolv Brand. 100/PK.
Catalog No. 50-014-52132 Supplier Microsolv Technology Corporation Supplier No. 9502S31CB
Caps, Screw Top, with Natural Silicone Rubber / White PTFE Pre-Slit Septa, bonded in knurled, polypropylene, Blue Caps. For use with 9-425mm thread, 12x32mm autosampler vials. MicroSolv Brand. 100/PK.