Learn More
Microsolv Technology Corporation Caps 2Ml Tan Sil/Ptfe Bl
Catalog No. 5001452106
Encompass_Preferred
Caps, Screw Top, with Tan Silicone Rubber / White PTFE Septa, bonded in knurled, polypropylene, Blue Caps. For use with 9-425mm thread, 12x32mm autosampler vials. AQ Brand. 100/PK.
Catalog No. 50-014-52106 Supplier Microsolv Technology Corporation Supplier No. 9502S11MB
Caps, Screw Top, with Tan Silicone Rubber / White PTFE Septa, bonded in knurled, polypropylene, Blue Caps. For use with 9-425mm thread, 12x32mm autosampler vials. AQ Brand. 100/PK.