Learn More
Microsolv Technology Corporation Caps 2Ml Tan Slit Sil/Ptfe Bl
Catalog No. 5001452133
Encompass_Preferred
Caps, Screw Top, with Tan Silicone Rubber / PTFE Pre-Slit Septa, bonded in knurled, polypropylene, Blue Caps. For use with 9-425mm thread, 12x32mm autosampler vials, AQ Brand. 100/PK.
Catalog No. 50-014-52133 Supplier Microsolv Technology Corporation Supplier No. 9502S31MB
Caps, Screw Top, with Tan Silicone Rubber / PTFE Pre-Slit Septa, bonded in knurled, polypropylene, Blue Caps. For use with 9-425mm thread, 12x32mm autosampler vials, AQ Brand. 100/PK.