Learn More
Alkali Scientific Caps for ASI™ 5mL False Bottom Analyzer Tubes with Threads, 16×97mm, Polypropylene (PP)
SDP
Catalog No. 5000663315
Encompass_Preferred
ASI™ caps are designed for use with 5 mL False Bottom Analyzer Tubes (16×97 mm). Manufactured from durable polypropylene (PP), these threaded caps provide a reliable and secure seal to help protect samples during storage, handling, and laboratory analysis. The precision threaded fit allows the cap to close tightly on compatible analyzer tubes, helping reduce the risk of leaks, evaporation, or contamination. Polypropylene construction offers excellent chemical resistance and durability for routine laboratory workflows. These caps are light yet sturdy, making them suitable for repeated handling in analytical, research, and diagnostic laboratory environments. Designed specifically for compatibility with ASI™ false bottom analyzer tubes, they help maintain sample integrity while supporting efficient and organized laboratory processing. Ideal for laboratories that require dependable tube sealing and consistent performance during sample preparation and analysis procedures.
Catalog No. 50-006-63315 Supplier Alkali Scientific Supplier No. FBATCAP
ASI™ caps are designed for use with 5 mL False Bottom Analyzer Tubes (16×97 mm). Manufactured from durable polypropylene (PP), these threaded caps provide a reliable and secure seal to help protect samples during storage, handling, and laboratory analysis. The precision threaded fit allows the cap to close tightly on compatible analyzer tubes, helping reduce the risk of leaks, evaporation, or contamination. Polypropylene construction offers excellent chemical resistance and durability for routine laboratory workflows. These caps are light yet sturdy, making them suitable for repeated handling in analytical, research, and diagnostic laboratory environments. Designed specifically for compatibility with ASI™ false bottom analyzer tubes, they help maintain sample integrity while supporting efficient and organized laboratory processing. Ideal for laboratories that require dependable tube sealing and consistent performance during sample preparation and analysis procedures.