Description
- Globe Scientific caps for automated systems fit several of the most popular platforms and fit seamlessly into capping processes.
- These cost-effective alternatives to OEM caps are manufactured at high standards to ensure efficient sample processing.
- Seamless cost-effective replacements for OEM caps
- Smooth operation in existing platforms
- Reliable sealing during high-speed transport and agitation
- Made from high-grade Polyethylene (PE) material
- Non-Sterile
Specifications
Specifications
| Type | Automated Sampling System Cap |
| For Use With (Equipment) | Beckman DXA5000 |
Note: All instruments and names of manufacturers mentioned herein are the trademarks of their respective companies and are used for reference purposes only. Globe Scientific LLC. Expressly disclaims any affiliation to them