Learn More
Microsolv Technology Corporation Caps Autosam9Mmsil/Ptfe Ps100P
Catalog No. 5001452123
Encompass_Preferred
Caps, Screw Top, with Silicone Rubber / PTFE Pre-Slit Septa, bonded in knurled, polypropylene, Blue Caps. For use with 9-425mm thread, 12x32mm autosampler vials. MicroSolv Brand. 100/PK
Catalog No. 50-014-52123 Supplier Microsolv Technology Corporation Supplier No. 9502S30CB
Caps, Screw Top, with Silicone Rubber / PTFE Pre-Slit Septa, bonded in knurled, polypropylene, Blue Caps. For use with 9-425mm thread, 12x32mm autosampler vials. MicroSolv Brand. 100/PK