Learn More
Microsolv Technology Corporation Caps Autosampler 9Mm Sil/Ptfe
Catalog No. 5001452097
Encompass_Preferred
Caps, Screw Top, with Silicone Rubber / PTFE, low durometer Septa, bonded in knurled, polypropylene, Blue Caps. For use with 9-425mm thread, 12x32mm autosampler vials. MicroSolv Brand. 100/PK.
Catalog No. 50-014-52097 Supplier Microsolv Technology Corporation Supplier No. 9502S10CB
Caps, Screw Top, with Silicone Rubber / PTFE, low durometer Septa, bonded in knurled, polypropylene, Blue Caps. For use with 9-425mm thread, 12x32mm autosampler vials. MicroSolv Brand. 100/PK.