Learn More
Microsolv Technology Corporation Caps, Crimp Top, with 0.05'' thick, Silicone Rubber / PTFE, ultra low bleed Septa, fitted in aluminum, Silver Caps. For use with 11mm Crimp or Snap Top, 12x32mm autosampler vials. 100/PK.
Catalog No. 502270371
Encompass_Preferred
Ultra low bleed Septa is a popular choice for GC and when siloxanes from silicone rubber may be a problem but you still want to use PTFE. Also used in HPLC when a Crimp Cap is required.
This item is not returnable. View return policy
Catalog No. 50-227-0371 Supplier Microsolv Technology Corporation Supplier No. 9502C41S
Ultra low bleed Septa is a popular choice for GC and when siloxanes from silicone rubber may be a problem but you still want to use PTFE. Also used in HPLC when a Crimp Cap is required.