Learn More
Microsolv Technology Corporation Caps, Crimp Top, with Soft-Guard, White Silicone / Red PTFE Septa, fitted in aluminum, Silver Caps. For use with 11mm Crimp or Snap Top, 12x32mm autosampler vials. 100/PK.
Catalog No. 502270425
Encompass_Preferred
Soft-Guard™ Septa provide easier penetration of autosampler needles and better resealing than other brands, for longer instrument up-time and fewer missed injections. Excellent for HPLC when Crimp Caps are required.
This item is not returnable. View return policy
Catalog No. 50-227-0425 Supplier Microsolv Technology Corporation Supplier No. 9502CE40TS
Soft-Guard™ Septa provide easier penetration of autosampler needles and better resealing than other brands, for longer instrument up-time and fewer missed injections. Excellent for HPLC when Crimp Caps are required.